Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HST4
Entrez Gene ID
851772
Gene Name
Gene Aliases
YDR191W
Chromosome Location
Chr.IV: 842337 - 843449 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180499.3 NP_010477.3 1 - 1 1056 69

View Full Product Details