Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS64
Entrez Gene ID
851781
Gene Name
Gene Aliases
FAR9, YDR200C
Chromosome Location
Chr.IV: 854279 - 856093 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180508.3 NP_010486.3 1 - 1 1748 75

View Full Product Details