Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCD6
Entrez Gene ID
851797
Gene Name
Gene Aliases
YDR211W
Chromosome Location
Chr.IV: 884727 - 886865 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180519.3 NP_010497.3 1 - 1 1852 68

View Full Product Details