Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFB1
Entrez Gene ID
851805
Gene Name
Gene Aliases
YDR219C
Chromosome Location
Chr.IV: 905455 - 906852 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180527.3 NP_010505.3 1 - 1 1248 76

View Full Product Details