Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNU56
Entrez Gene ID
851827
Gene Name
Gene Aliases
MUD10, YDR240C
Chromosome Location
Chr.IV: 943674 - 945152 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180548.1 NP_010526.1 1 - 1 1046 91

View Full Product Details