Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRS23
Entrez Gene ID
851833
Gene Name
Gene Aliases
YDR246W
Chromosome Location
Chr.IV: 954288 - 954947 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180554.3 NP_010532.3 1 - 1 451 71

View Full Product Details