Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YAP6
Entrez Gene ID
851846
Gene Name
Gene Aliases
HAL7, YDR259C
Chromosome Location
Chr.IV: 974631 - 975782 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180567.3 NP_010545.3 1 - 1 651 82

View Full Product Details