Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AKR1
Entrez Gene ID
851857
Gene Name
Gene Aliases
YDR264C
Chromosome Location
Chr.IV: 996029 - 998323 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180572.1 NP_010550.1 1 - 1 2052 74

View Full Product Details