Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUR2
Entrez Gene ID
851891
Gene Name
Gene Aliases
SYR2, YDR297W
Chromosome Location
Chr.IV: 1056551 - 1057600 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180605.1 NP_010583.1 1 - 1 756 79

View Full Product Details