Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRS1
Entrez Gene ID
851942
Gene Name
Gene Aliases
YDR341C
Chromosome Location
Chr.IV: 1151803 - 1153626 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180649.3 NP_010628.3 1 - 1 1749 85

View Full Product Details