Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRQ1
Entrez Gene ID
851979
Gene Name
Gene Aliases
YDR373W
Chromosome Location
Chr.IV: 1222759 - 1223331 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180681.3 NP_010661.3 1 - 1 57 72

View Full Product Details