Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NKP1
Entrez Gene ID
851991
Gene Name
Gene Aliases
YDR383C
Chromosome Location
Chr.IV: 1239960 - 1240676 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180691.3 NP_010671.4 1 - 1 646 72

View Full Product Details