Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPB7
Entrez Gene ID
852013
Gene Name
Gene Aliases
YDR404C
Chromosome Location
Chr.IV: 1276654 - 1277169 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180712.3 NP_010692.3 1 - 1 447 93

View Full Product Details