Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEH2
Entrez Gene ID
852069
Gene Name
Gene Aliases
YDR458C
Chromosome Location
Chr.IV: 1380055 - 1382046 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180766.1 NP_010746.1 1 - 1 1851 98

View Full Product Details