Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPP41
Entrez Gene ID
852075
Gene Name
Gene Aliases
YDR464W
Chromosome Location
Chr.IV: 1388872 - 1393179 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180772.3 NP_010752.3 1 - 1 4242 97

View Full Product Details