Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIG2
Entrez Gene ID
852091
Gene Name
Gene Aliases
RST2, YDR480W
Chromosome Location
Chr.IV: 1417399 - 1418370 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180788.1 NP_010768.1 1 - 1 643 76

View Full Product Details