Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS60
Entrez Gene ID
852097
Gene Name
Gene Aliases
CHM5, MOS10, YDR486C
Chromosome Location
Chr.IV: 1427431 - 1428120 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180794.3 NP_010774.4 1 - 1 551 71

View Full Product Details