Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIN4
Entrez Gene ID
852119
Gene Name
Gene Aliases
ERC47, YDR507C
Chromosome Location
Chr.IV: 1462358 - 1465786 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180815.3 NP_010795.3 1 - 1 3350 89

View Full Product Details