Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
URA3
Entrez Gene ID
856692
Gene Name
Gene Aliases
YEL021W
Chromosome Location
Chr.V: 116167 - 116970 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178836.3 NP_010893.3 1 - 1 749 77

View Full Product Details