Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HXT13
Entrez Gene ID
856640
Gene Name
Gene Aliases
HLT1, YEL069C
Chromosome Location
Chr.V: 21537 - 23231 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178884.1 NP_010845.1 1 - 1 47 99

View Full Product Details