Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ICL1
Entrez Gene ID
856794
Gene Name
Gene Aliases
YER065C
Chromosome Location
Chr.V: 285241 - 286914 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178956.3 NP_010987.3 1 - 1 1552 88

View Full Product Details