Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALD5
Entrez Gene ID
856804
Gene Name
Gene Aliases
YER073W
Chromosome Location
Chr.V: 304030 - 305592 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178964.2 NP_010996.2 1 - 1 1449 96

View Full Product Details