Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIM9
Entrez Gene ID
856813
Gene Name
Gene Aliases
FMP29, YER080W
Chromosome Location
Chr.V: 319963 - 321846 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178971.1 NP_011003.1 1 - 1 1748 74

View Full Product Details