Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRP2
Entrez Gene ID
856824
Gene Name
Gene Aliases
YER090W
Chromosome Location
Chr.V: 337949 - 339472 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178981.1 NP_011014.1 1 - 1 1446 104

View Full Product Details