Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YCK3
Entrez Gene ID
856860
Gene Name
Gene Aliases
CKI3, YER123W
Chromosome Location
Chr.V: 404813 - 406387 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179013.1 NP_011049.2 1 - 1 1348 91

View Full Product Details