Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP12
Entrez Gene ID
850532
Gene Name
Gene Aliases
GLP1, HOR5, YFL014W
Chromosome Location
Chr.VI: 107256 - 107585 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179952.1 NP_116640.1 1 - 1 248 79

View Full Product Details