Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FET5
Entrez Gene ID
850502
Gene Name
Gene Aliases
YFL041W
Chromosome Location
Chr.VI: 49139 - 51007 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179925.1 NP_116612.1 1 - 1 1750 108

View Full Product Details