Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YFH7
Entrez Gene ID
850558
Gene Name
Gene Aliases
AIM12, YFR007W
Chromosome Location
Chr.VI: 159299 - 160360 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179972.1 NP_116662.1 1 - 1 943 68

View Full Product Details