Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ULI1
Entrez Gene ID
850583
Gene Name
Gene Aliases
YFR026C
Chromosome Location
Chr.VI: 205748 - 206257 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179991.3 NP_116682.3 1 - 1 352 62

View Full Product Details