Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPN14
Entrez Gene ID
852880
Gene Name
Gene Aliases
YGL004C
Chromosome Location
Chr.VII: 490703 - 491956 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180869.3 NP_011511.3 1 - 1 1154 91

View Full Product Details