Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUT1
Entrez Gene ID
852714
Gene Name
Gene Aliases
YGL162W
Chromosome Location
Chr.VII: 198138 - 199037 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181027.1 NP_011353.1 1 - 1 752 103

View Full Product Details