Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIF3
Entrez Gene ID
852651
Gene Name
Gene Aliases
YGL221C
Chromosome Location
Chr.VII: 81426 - 82292 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181086.1 NP_011294.1 1 - 1 47 77

View Full Product Details