Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAI1
Entrez Gene ID
852646
Gene Name
Gene Aliases
YGL246C
Chromosome Location
Chr.VII: 37617 - 38780 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181112.1 NP_011268.1 1 - 1 947 102

View Full Product Details