Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NMA2
Entrez Gene ID
852893
Gene Name
Gene Aliases
YGR010W
Chromosome Location
Chr.VII: 511545 - 512732 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181139.1 NP_011524.1 1 - 1 947 86

View Full Product Details