Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNU71
Entrez Gene ID
852896
Gene Name
Gene Aliases
YGR013W
Chromosome Location
Chr.VII: 514554 - 516416 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181142.1 NP_011527.1 1 - 1 1752 66

View Full Product Details