Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCM4
Entrez Gene ID
852940
Gene Name
Gene Aliases
YGR049W
Chromosome Location
Chr.VII: 591314 - 591877 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181178.1 NP_011563.1 1 - 1 450 90

View Full Product Details