Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRP13
Entrez Gene ID
852975
Gene Name
Gene Aliases
YGR084C, mS44
Chromosome Location
Chr.VII: 647123 - 648142 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181213.3 NP_011598.3 1 - 1 951 81

View Full Product Details