Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLB6
Entrez Gene ID
853003
Gene Name
Gene Aliases
YGR109C
Chromosome Location
Chr.VII: 705359 - 706501 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181238.3 NP_011623.3 1 - 1 951 96

View Full Product Details