Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NSR1
Entrez Gene ID
853064
Gene Name
Gene Aliases
SHE5, YGR159C
Chromosome Location
Chr.VII: 806412 - 807656 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181288.1 NP_011675.1 1 - 1 947 84

View Full Product Details