Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YIP1
Entrez Gene ID
853082
Gene Name
Gene Aliases
YGR172C
Chromosome Location
Chr.VII: 842845 - 843591 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181301.3 NP_011688.3 1 - 1 548 82

View Full Product Details