Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBG2
Entrez Gene ID
853083
Gene Name
Gene Aliases
GIR1, YGR173W
Chromosome Location
Chr.VII: 843854 - 844960 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181302.1 NP_011689.1 1 - 1 1055 83

View Full Product Details