Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XKS1
Entrez Gene ID
853108
Gene Name
Gene Aliases
YGR194C
Chromosome Location
Chr.VII: 886073 - 887875 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181323.3 NP_011710.3 1 - 1 1746 92

View Full Product Details