Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AMA1
Entrez Gene ID
853140
Gene Name
Gene Aliases
SPO70, YGR225W
Chromosome Location
Chr.VII: 945145 - 947019 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181354.3 NP_011741.5 1 - 2 1184 65

View Full Product Details