Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDA1
Entrez Gene ID
853160
Gene Name
Gene Aliases
YGR245C
Chromosome Location
Chr.VII: 979765 - 982068 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181374.3 NP_011761.3 1 - 1 2145 90

View Full Product Details