Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTM1
Entrez Gene ID
853173
Gene Name
Gene Aliases
YGR257C
Chromosome Location
Chr.VII: 1006205 - 1007305 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181386.3 NP_011773.3 1 - 1 854 69

View Full Product Details