Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HUA1
Entrez Gene ID
853185
Gene Name
Gene Aliases
YGR268C
Chromosome Location
Chr.VII: 1026057 - 1026653 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181397.3 NP_011784.3 1 - 1 444 96

View Full Product Details