Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHU1
Entrez Gene ID
856383
Gene Name
Gene Aliases
YHL006C
Chromosome Location
Chr.VIII: 98343 - 98795 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179086.1 NP_011857.2 1 - 1 346 100

View Full Product Details