Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WSC4
Entrez Gene ID
856357
Gene Name
Gene Aliases
YFW1, YHC8, YHL028W
Chromosome Location
Chr.VIII: 48763 - 50580 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179108.1 NP_011835.1 1 - 1 1744 95

View Full Product Details