Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COS8
Entrez Gene ID
856337
Gene Name
Gene Aliases
YHL048W
Chromosome Location
Chr.VIII: 6401 - 7546 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179128.1 NP_011815.1 1 - 1 1046 85

View Full Product Details