Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FSH1
Entrez Gene ID
856445
Gene Name
Gene Aliases
YHR049W
Chromosome Location
Chr.VIII: 206462 - 207193 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179179.1 NP_011915.1 1 - 1 649 70

View Full Product Details